Learn More
Selleck Chemical LLC PIM447 (LGH447) 2mg 1210608-43-7(freebase)
SDP
Supplier:  Selleck Chemical LLC S79852MG
Encompass_Preferred
PIM447 (LGH447) is a novel pan-PIM kinase inhibitor with Ki values of 6 pM, 18 pM, 9 pM for PIM1, PIM2, PIM3 respectively. It also inhibits GSK3?, PKN1, and PKC?, but at a significantly lower potency with IC50 between 1 and 5 ?M (>105-fold differential relative to the Ki on PIMs). PIM447 induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5512