Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pimafucin (Pimaricin in A 2.5% Sterile Water Suspension), Thermo Scientific™

Catalog No. AC131220200
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC131220200 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC131220200 Supplier Thermo Scientific Chemicals Supplier No. 131220200

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-93-8
Molecular Formula C33H47NO13
Molecular Weight (g/mol) 665.733
InChI Key NCXMLFZGDNKEPB-UWSQUGIJSA-N
Synonym natamycin
PubChem CID 129317863
SMILES CC1CC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(CC3C(O3)C=CC(=O)O1)O)O)O)C(=O)O)OC4C(C(C(C(O4)C)O)N)O

Specifications

Merck Index 15, 6509
Formula Weight 665.72
Quantity 20 mL
Chemical Name or Material Pimafucin (Pimaricin in a 2.5% sterile water suspension)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.