Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pimaricin, 2.5% aqueous suspension, MP Biomedicals™

Catalog No. ICN15625420 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15625420 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15625420 Supplier MP Biomedicals, Inc Supplier No. 0215625420
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An amphoteric macrolide antifungal antibiotic used for studying lipid bilayer dynamics and as a fungicide in agar media.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-93-8
Molecular Formula C33H47NO13
Molecular Weight (g/mol) 665.733
InChI Key NCXMLFZGDNKEPB-UWSQUGIJSA-N
Synonym Antibiotic A-5283, Pimafucin
PubChem CID 129317863
SMILES CC1CC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(CC3C(O3)C=CC(=O)O1)O)O)O)C(=O)O)OC4C(C(C(C(O4)C)O)N)O

Specifications

Type Pimaricin
Physical Form Suspension
Color Cream
Quantity 20 mL
Formula Weight 665.75
Grade Not Available
Chemical Name or Material Pimaricin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.