Learn More
Medchemexpress LLC HY-12042 10mg , Pimasertib CAS:1236699-92-5 Purity:>98%
SDP
Catalog No. 502259974
Encompass_Preferred
HY-12042 10mg Pimasertib CAS: 1236699-92-5 Pimasertib (AS703026) is a highly selective, ATP non-competitive allosteric orally available MEK1/2 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9974 Supplier Medchemexpress LLC Supplier No. HY1204210MG
HY-12042 10mg Pimasertib CAS: 1236699-92-5 Pimasertib (AS703026) is a highly selective, ATP non-competitive allosteric orally available MEK1/2 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.