Learn More
Medchemexpress LLC N-[(2S)-2,3-dihydroxypropyl]-3-(2-fluoro-4-iodoanilino)pyridine-4-carboxamide | 1236699-92-5 | 5 MG
SDP
Catalog No. 502080940
Encompass_Preferred
Pimasertib is a selective, ATP non-competitive allosteric inhibitor of MEK1/2 used in preclinical research to investigate MAPK/ERK signaling and antiproliferative effects. Supplied in a small, precise quantity for laboratory assays.
  • Selective MEK1/2 inhibition for pathway-targeted studies
  • ATP non-competitive allosteric mechanism of action
  • Supplied as a 5 mg solid for small-scale experiments
  • Well characterized by CAS and chemical identifiers
  • Suitable for cell-based and biochemical assays
Catalog No. 50-208-0940 Supplier Medchemexpress LLC Supplier No. HY120425MG
Pimasertib is a selective, ATP non-competitive allosteric inhibitor of MEK1/2 used in preclinical research to investigate MAPK/ERK signaling and antiproliferative effects. Supplied in a small, precise quantity for laboratory assays.
  • Selective MEK1/2 inhibition for pathway-targeted studies
  • ATP non-competitive allosteric mechanism of action
  • Supplied as a 5 mg solid for small-scale experiments
  • Well characterized by CAS and chemical identifiers
  • Suitable for cell-based and biochemical assays