Learn More
Medchemexpress LLC HY-14557 25mg Medchemexpress, Pimavanserin CAS:706779-91-1 Purity:>98%
SDP
Catalog No. 501969910
Encompass_Preferred
Medchemexpress, HY-14557 25mg Pimavanserin CAS:706779-91-1 Pimavanserin is a selective inverse agonist of the 5-HT2A receptor with pIC50 and pKd of 8.73 and 9.3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9910 Supplier Medchemexpress LLC Supplier No. HY1455725MG
Medchemexpress, HY-14557 25mg Pimavanserin CAS:706779-91-1 Pimavanserin is a selective inverse agonist of the 5-HT2A receptor with pIC50 and pKd of 8.73 and 9.3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.