Learn More
Apexbio Technology LLC Pimavanserin 706779-91-1 5mg
SDP
Catalog No. 502240792
Encompass_Preferred
Pimavanserin (CAS 706779-91-1) is a small-molecule inverse agonist that selectively binds to the serotonin 5-HT2A receptor In cell-based functional assays it exhibits high potency with a mean pIC50 value of 8 7 Pimavanserin is widely utilized in research to investigate serotonergic signaling pathways the pharmacology of 5-HT2A receptor modulation and potential therapeutic approaches for neuropsychiatric disorders involving dysregulated serotonin function
This item is not returnable. View return policy
Catalog No. 50-224-0792 Supplier Apexbio Technology LLC Supplier No. B80195
Pimavanserin (CAS 706779-91-1) is a small-molecule inverse agonist that selectively binds to the serotonin 5-HT2A receptor In cell-based functional assays it exhibits high potency with a mean pIC50 value of 8 7 Pimavanserin is widely utilized in research to investigate serotonergic signaling pathways the pharmacology of 5-HT2A receptor modulation and potential therapeutic approaches for neuropsychiatric disorders involving dysregulated serotonin function