Learn More
Medchemexpress LLC HY-14557A 25mg Medchemexpress, Pimavanserin tartrate CAS:706782-28-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14557A25MG
Encompass_Preferred
Medchemexpress, HY-14557A 25mg Pimavanserin tartrate CAS:706782-28-7 Pimavanserin tartrate (ACP-103) is a potent 5-HT 2A receptor inverse agonist with pIC50 and pKi of 8.73 and 9.3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9911