Learn More
Selleck Chemical LLC Pimavanserin tartrate
SDP
Catalog No. 5000001903
Encompass_Preferred
Pimavanserin tartrate(ACP-103) is a potent and selective serotonin 5-HT2A inverse agonist with pIC50 of 8 73 used to treat psychosis associated with Parkinson s disease
Catalog No. 50-000-01903 Supplier Selleck Chemical LLC Supplier No. S818310MM/1ML
Pimavanserin tartrate(ACP-103) is a potent and selective serotonin 5-HT2A inverse agonist with pIC50 of 8 73 used to treat psychosis associated with Parkinson s disease