Learn More
Cayman Chemical Pimavanserin, Purity: ≥98%, CAS: 706779-91-1, 50 mg
SDP
Catalog No. 502994632
Encompass_Preferred
A 5-HT2A inverse agonist (IC50 = 1.86 nM; Ki = 0.5 nM); selective for 5-HT2A over a panel of 65 ion channels, enzymes, and receptors (Kis = >100 nM); reduces DOI-induced head-twitch behavior and prepulse inhibition deficits in rats at doses of 3 mg/kg, p.o. and 1-10 mg/kg, s.c., respectively; exhibits antipsychotic-like activity, reducing (+)-MK-801-induced hyperactivity in mice; acts synergistically with haloperidol or risperidone to suppress (+)-MK-801-induced hyperactivity and attenuates haloperidol- and risperidone-induced catalepsy in rats
Catalog No. 50-299-4632 Supplier Cayman Chemical Supplier No. 2346250 MG
A 5-HT2A inverse agonist (IC50 = 1.86 nM; Ki = 0.5 nM); selective for 5-HT2A over a panel of 65 ion channels, enzymes, and receptors (Kis = >100 nM); reduces DOI-induced head-twitch behavior and prepulse inhibition deficits in rats at doses of 3 mg/kg, p.o. and 1-10 mg/kg, s.c., respectively; exhibits antipsychotic-like activity, reducing (+)-MK-801-induced hyperactivity in mice; acts synergistically with haloperidol or risperidone to suppress (+)-MK-801-induced hyperactivity and attenuates haloperidol- and risperidone-induced catalepsy in rats