Learn More
TARGETMOL CHEMICALS INC PIMECROLIMUS 100MG
SDP
Catalog No. 502577072
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pimecrolimus (SDZ-ASM 981) a calcineurin inhibitor immunosuppressant binds to the receptor macrophilin-12 (FKBP-12) forming a complex that blocks the calcium-dependent signal transduction cascade mediated by calcineurin. purity: 99%
Catalog No. 50-257-7072 Supplier TARGETMOL CHEMICALS INC Supplier No. T2648100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pimecrolimus (SDZ-ASM 981) a calcineurin inhibitor immunosuppressant binds to the receptor macrophilin-12 (FKBP-12) forming a complex that blocks the calcium-dependent signal transduction cascade mediated by calcineurin. purity: 99%