Learn More
AdipoGen Pimecrolimus (50 mg)
SDP
Catalog No. 502054625
Encompass_Preferred
Chemical. CAS: 137071-32-0. Formula: C43H68ClNO11. MW: 810.45. Synthetic. Pimecrolimus is a immunosuppressive natural ascomycin macrolactam that binds to macrophilin-12 (FKBP-12) and inhibits calcineurin as well as prolyl isomerase (rotamase). Presumably through these actions, it blocks the activation of T cells by allogeneic dendritic cells (IC50=0.55nM) without affecting dendritic cells. Moreover, pimecrolimus suppresses the generation of pro-inflammatory cytokines by T cells, the release of pre-formed inflammatory mediators from mast cells and the activation of eosinophils. These effects support the use of pimecrolimus in countering inflammatory skin diseases, such as atopic dermatitis (eczema) and psoriasis.
Catalog No. 50-205-4625 Supplier AdipoGen Supplier No. CDXP0598M050
Chemical. CAS: 137071-32-0. Formula: C43H68ClNO11. MW: 810.45. Synthetic. Pimecrolimus is a immunosuppressive natural ascomycin macrolactam that binds to macrophilin-12 (FKBP-12) and inhibits calcineurin as well as prolyl isomerase (rotamase). Presumably through these actions, it blocks the activation of T cells by allogeneic dendritic cells (IC50=0.55nM) without affecting dendritic cells. Moreover, pimecrolimus suppresses the generation of pro-inflammatory cytokines by T cells, the release of pre-formed inflammatory mediators from mast cells and the activation of eosinophils. These effects support the use of pimecrolimus in countering inflammatory skin diseases, such as atopic dermatitis (eczema) and psoriasis.