Learn More
Cayman Chemical PImecrolImus 50mg
SDP
Catalog No. 502952541
Encompass_Preferred
A macrolactam that binds to macrophilin-12 and inhibits calcineurin as well as prolyl isomerase; the activation of T cells by allogeneic dendritic cells (IC50 = 0.55 nM) and suppresses the generation of pro-inflammatory cytokines; used in countering inflammatory skin diseases, such as eczema and psoriasis
Catalog No. 50-295-2541 Supplier Cayman Chemical Supplier No. 1310750 MG
A macrolactam that binds to macrophilin-12 and inhibits calcineurin as well as prolyl isomerase; the activation of T cells by allogeneic dendritic cells (IC50 = 0.55 nM) and suppresses the generation of pro-inflammatory cytokines; used in countering inflammatory skin diseases, such as eczema and psoriasis