Learn More
Medchemexpress LLC Pimelic acid | 111-16-0 | MFCD00004425 | ≥98.0% | 160.17 g/mol | C7H12O4 | 5g
SDP
Catalog No. 5000266946
Encompass_Preferred
Pimelic acid is the organic compound and its derivatives are involved in the biosynthesis of the amino acid called lysine Pimelic acid can be used in keratoconus research[1]
Catalog No. 50-002-66946 Supplier Medchemexpress LLC Supplier No. HYY11395G
Pimelic acid is the organic compound and its derivatives are involved in the biosynthesis of the amino acid called lysine Pimelic acid can be used in keratoconus research[1]