Learn More
Selleck Chemical LLC Pimelic acid S3145-100mg
SDP
Catalog No. 503110031
Encompass_Preferred
Pimelic acid (Heptanedioic acid 1 5-Pentanedicarboxylic acid 1 7-Heptanedioic acid) originating from fatty acid synthesis pathway is a bona fide precursor of biotin in B subtilis
Catalog No. 50-311-0031 Supplier Selleck Chemical LLC Supplier No. S3145100MG
Pimelic acid (Heptanedioic acid 1 5-Pentanedicarboxylic acid 1 7-Heptanedioic acid) originating from fatty acid synthesis pathway is a bona fide precursor of biotin in B subtilis