Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pimelic acid

Catalog No. P45001100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P45001100G 100 g
P4500125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P45001100G Supplier Sigma Aldrich Supplier No. P45001100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pimelic acid

Specifications

CAS 111-16-0
Melting Point 103°C to 105°C (lit.)
Boiling Point 212°C (10 mmHg)
Molecular Formula C7H12O4
Linear Formula HO2C(CH2)5 CO2H
MDL Number MFCD00004425
Quantity 100 g
Synonym Heptanedioic acid
Molecular Weight (g/mol) 160.17
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.