Learn More
Selleck Chemical LLC Pimethixene E4862-5MG
SDP
Catalog No. 5000043304
Encompass_Preferred
Pimethixene(Calmixen Pimetixene) is a highly potent and non-selective antagonist of the 5-HT2B receptor with a pKi of 10 44 It also functions as an antihistamine and anticholinergic agent and is a highly potent multi-target antagonist with antagonistic effects on multiple receptors
Catalog No. 50-000-43304 Supplier Selleck Chemical LLC Supplier No. E48625MG
Pimethixene(Calmixen Pimetixene) is a highly potent and non-selective antagonist of the 5-HT2B receptor with a pKi of 10 44 It also functions as an antihistamine and anticholinergic agent and is a highly potent multi-target antagonist with antagonistic effects on multiple receptors