Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pimethixene maleate | 13187-06-9 | 99.7% | 5 MG
SDP

Catalog No. 5000413371
Encompass_Preferred

Pimethixene maleate is an antihistamine and antiserotonergic compound that acts as an antimigraine agent. It is a highly potent antagonist of 5-HT1A, 5-HT2A, 5-HT2B, 5-HT2C, histamine H1, dopamine D2 and D4.4, as well as muscarinic M1 and M2 receptors. The pKis for these receptors are 7.63, 10.22, 10.44, 8.42, 10.14, 8.19, 7.54, 8.61, and 9.38, respectively.

  • Antihistamine and antiserotonergic compound.
  • Acts as an antimigraine agent.
  • Highly potent antagonist of multiple receptors including 5-HT1A, 5-HT2A, 5-HT2B, 5-HT2C, histamine H1, dopamine D2 and D4.4, and muscarinic M1 and M2.
  • For research use only.

Catalog No. 50-004-13371 Supplier Medchemexpress LLC Supplier No. HYB1101A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pimethixene maleate is an antihistamine and antiserotonergic compound that acts as an antimigraine agent. It is a highly potent antagonist of 5-HT1A, 5-HT2A, 5-HT2B, 5-HT2C, histamine H1, dopamine D2 and D4.4, as well as muscarinic M1 and M2 receptors. The pKis for these receptors are 7.63, 10.22, 10.44, 8.42, 10.14, 8.19, 7.54, 8.61, and 9.38, respectively.

  • Antihistamine and antiserotonergic compound.
  • Acts as an antimigraine agent.
  • Highly potent antagonist of multiple receptors including 5-HT1A, 5-HT2A, 5-HT2B, 5-HT2C, histamine H1, dopamine D2 and D4.4, and muscarinic M1 and M2.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.