Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pimethixene maleate | 13187-06-9 | MFCD00079242 | 99.7% | 1 ML
SDP

Catalog No. 5000413326
Encompass_Preferred

Pimethixene maleate is an antihistamine and antiserotonergic compound that functions as an antimigraine agent. It is a potent antagonist for several receptors, including 5-HT1A, 5-HT2A, 5-HT2B, 5-HT2C, histamine H1, dopamine D2 and D4.4, as well as muscarinic M1 and M2 receptors. It is intended for research use only.

  • Antihistamine and antiserotonergic compound
  • Acts as an antimigraine agent
  • Potent antagonist of various receptors (5-HT1A, 5-HT2A, 5-HT2B, 5-HT2C, histamine H1, dopamine D2 and D4.4, muscarinic M1 and M2)
  • Molecular weight: 409.50
  • Formula: C23H23NO4S
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-004-13326 Supplier Medchemexpress LLC Supplier No. HYB1101A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pimethixene maleate is an antihistamine and antiserotonergic compound that functions as an antimigraine agent. It is a potent antagonist for several receptors, including 5-HT1A, 5-HT2A, 5-HT2B, 5-HT2C, histamine H1, dopamine D2 and D4.4, as well as muscarinic M1 and M2 receptors. It is intended for research use only.

  • Antihistamine and antiserotonergic compound
  • Acts as an antimigraine agent
  • Potent antagonist of various receptors (5-HT1A, 5-HT2A, 5-HT2B, 5-HT2C, histamine H1, dopamine D2 and D4.4, muscarinic M1 and M2)
  • Molecular weight: 409.50
  • Formula: C23H23NO4S
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.