Learn More
Selleck Chemical LLC Pimitespib TAS-116-5mg
SDP
Catalog No. 502389582
Encompass_Preferred
Pimitespib (TAS-116) is a novel, small-molecule HSP90 inhibitor which inhibits geldanamycin-FITC binding to HSP90 proteins with Ki values of 34.7 nmol/L, 21.3 nmol/L, >50,000 nmol/L, and >50,000 nmol/L for HSP90α, HSP90β, GRP94, and TRAP1, respectively. Furthermore, TAS-116 does not inhibit other ATPases such as HSP70 (IC50 >200 μmol/L).
Catalog No. 50-238-9582 Supplier Selleck Chemical LLC Supplier No. S77165MG
Pimitespib (TAS-116) is a novel, small-molecule HSP90 inhibitor which inhibits geldanamycin-FITC binding to HSP90 proteins with Ki values of 34.7 nmol/L, 21.3 nmol/L, >50,000 nmol/L, and >50,000 nmol/L for HSP90α, HSP90β, GRP94, and TRAP1, respectively. Furthermore, TAS-116 does not inhibit other ATPases such as HSP70 (IC50 >200 μmol/L).