Learn More
Apexbio Technology LLC Pimobendan 74150-27-9 200mg
SDP
Catalog No. 5000095384
Encompass_Preferred
Pimobendan (CAS 74150-27-9) is a small molecule that acts as a selective phosphodiesterase 3 (PDE3) inhibitor exhibiting an IC50 of 0 32 M By inhibiting PDE3 activity Pimobendan modulates intracellular cyclic nucleotide levels thereby influencing cellular signaling pathways involved in cardiovascular function This compound is utilized in biomedical research to investigate PDE3-mediated mechanisms and their implications in cardiac physiology and related disease models
Catalog No. 50-000-95384 Supplier Apexbio Technology LLC Supplier No. A4340200
Pimobendan (CAS 74150-27-9) is a small molecule that acts as a selective phosphodiesterase 3 (PDE3) inhibitor exhibiting an IC50 of 0 32 M By inhibiting PDE3 activity Pimobendan modulates intracellular cyclic nucleotide levels thereby influencing cellular signaling pathways involved in cardiovascular function This compound is utilized in biomedical research to investigate PDE3-mediated mechanisms and their implications in cardiac physiology and related disease models