Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pimobendan-d3 | 74150-27-9 | 99.4% | 337.39 | C19H15D3N4O2 | 10 MG
SDP

Catalog No. 5000298883
Encompass_Preferred

Pimobendan-d3 is a deuterium-labeled isotopologue of pimobendan supplied as a stable-isotope reference standard for analytical and research use. It is intended for LC-MS internal standard and tracer studies and shares the pharmacological annotation of the parent compound (pimobendan), a selective PDE3 inhibitor (IC50 = 0.32 μM).

  • Deuterium-labeled internal standard for LC-MS and tracer studies.
  • High purity (99.43%).
  • Molecular weight 337.39 Da.
  • CAS number 74150-27-9 for unambiguous identification.
  • Available in small vial sizes, including 10 mg, for analytical workflows.

Catalog No. 50-002-98883 Supplier Medchemexpress LLC Supplier No. HYB0204S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pimobendan-d3 is a deuterium-labeled isotopologue of pimobendan supplied as a stable-isotope reference standard for analytical and research use. It is intended for LC-MS internal standard and tracer studies and shares the pharmacological annotation of the parent compound (pimobendan), a selective PDE3 inhibitor (IC50 = 0.32 μM).

  • Deuterium-labeled internal standard for LC-MS and tracer studies.
  • High purity (99.43%).
  • Molecular weight 337.39 Da.
  • CAS number 74150-27-9 for unambiguous identification.
  • Available in small vial sizes, including 10 mg, for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.