Learn More
Medchemexpress LLC Pimobendan-d3 | 74150-27-9 | 99.4% | 50 MG
SDP
Catalog No. 5000405812
Encompass_Preferred
Pimobendan-d3 (UD-CG115-d3) is deuterium labeled Pimobendan. It is a selective inhibitor of PDE3 with an IC50 of 0.32 μM. Deuterium labeled compounds are used as tracers for quantitation during drug development due to their potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-004-05812 Supplier Medchemexpress LLC Supplier No. HYB0204S150MG
Pimobendan-d3 (UD-CG115-d3) is deuterium labeled Pimobendan. It is a selective inhibitor of PDE3 with an IC50 of 0.32 μM. Deuterium labeled compounds are used as tracers for quantitation during drug development due to their potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only