Learn More
Abcam Pimobendan (Vetmedin), Ca2+ channel sensitizer, 5MG
Catalog No. 502706136
Encompass_Preferred
MW 334.4 Da, Purity >99%. Ca2+ channel sensitizer. Selective PDE3 inhibitor. Inhibits platelet aggregation. Shows antithrombotic, antihypertensive, potent inotropic and vasodilatory effects *in vivo.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6136 Supplier Abcam Supplier No. AB1430195MG
MW 334.4 Da, Purity >99%. Ca2+ channel sensitizer. Selective PDE3 inhibitor. Inhibits platelet aggregation. Shows antithrombotic, antihypertensive, potent inotropic and vasodilatory effects *in vivo.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement