Learn More
Medchemexpress LLC HY-12353A 10mg Medchemexpress, Pimodivir CAS:1629869-44-8 Purity:>98%
SDP
Catalog No. 502028875
Encompass_Preferred
Medchemexpress, HY-12353A 10mg Pimodivir CAS:1629869-44-8 Pimodivir (VX-787) is an orally bioavailable inhibitor of influenza A virus polymerases through interaction with the viral PB2 subunit. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8875 Supplier Medchemexpress LLC Supplier No. HY12353A10MG
Medchemexpress, HY-12353A 10mg Pimodivir CAS:1629869-44-8 Pimodivir (VX-787) is an orally bioavailable inhibitor of influenza A virus polymerases through interaction with the viral PB2 subunit. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.