Learn More
Cayman Chemical Pimonidazole, Purity: ≥98%, CAS: 70132-50-2, 100 mg
SDP
Catalog No. 502896748
Encompass_Preferred
A small molecule radiosensitizer that has proven to be an effective and nontoxic immunochemical hypoxia marker for human squamous cell carcinomas of the cervix, head and neck
Catalog No. 50-289-6748 Supplier Cayman Chemical Supplier No. 89130100 MG
A small molecule radiosensitizer that has proven to be an effective and nontoxic immunochemical hypoxia marker for human squamous cell carcinomas of the cervix, head and neck