Learn More
Cayman Chemical Pimonidazole, Purity: ≥98%, CAS: 70132-50-2, 10 mg
SDP
Catalog No. 502892769
Encompass_Preferred
A small molecule radiosensitizer that has proven to be an effective and nontoxic immunochemical hypoxia marker for human squamous cell carcinomas of the cervix, head and neck
Catalog No. 50-289-2769 Supplier Cayman Chemical Supplier No. 8913010 MG
A small molecule radiosensitizer that has proven to be an effective and nontoxic immunochemical hypoxia marker for human squamous cell carcinomas of the cervix, head and neck