Learn More
Cayman Chemical PImonIdazole 50mg
SDP
Catalog No. 502892411
Encompass_Preferred
A small molecule radiosensitizer that has proven to be an effective and nontoxic immunochemical hypoxia marker for human squamous cell carcinomas of the cervix, head and neck
Catalog No. 50-289-2411 Supplier Cayman Chemical Supplier No. 8913050 MG
A small molecule radiosensitizer that has proven to be an effective and nontoxic immunochemical hypoxia marker for human squamous cell carcinomas of the cervix, head and neck