Learn More
Medchemexpress LLC Pimonidazole | 70132-50-2 | MFCD00867974 | 99.3% | 254.29 g/mol | C11H18N4O3 | 5 MG
SDP
Catalog No. 5000243845
Encompass_Preferred
Pimonidazole is a nitroimidazole-based hypoxia marker used in research to detect and quantify low-oxygen regions in cells and tissues. After bioreduction under hypoxic conditions, it forms covalent adducts with macromolecules that can be detected by immunohistochemistry or other analytical methods, enabling both qualitative and quantitative assessment of tumor and tissue hypoxia.
  • Selective hypoxia marker for cellular and tissue studies.
  • Forms stable protein adducts after bioreduction in low-oxygen environments.
  • Compatible with immunohistochemistry and hypoxia assay workflows.
  • Supplied as high-purity material suitable for analytical and research use.
  • Useful for both qualitative and quantitative hypoxia assessment.
Catalog No. 50-002-43845 Supplier Medchemexpress LLC Supplier No. HY105129A5MG
Pimonidazole is a nitroimidazole-based hypoxia marker used in research to detect and quantify low-oxygen regions in cells and tissues. After bioreduction under hypoxic conditions, it forms covalent adducts with macromolecules that can be detected by immunohistochemistry or other analytical methods, enabling both qualitative and quantitative assessment of tumor and tissue hypoxia.
  • Selective hypoxia marker for cellular and tissue studies.
  • Forms stable protein adducts after bioreduction in low-oxygen environments.
  • Compatible with immunohistochemistry and hypoxia assay workflows.
  • Supplied as high-purity material suitable for analytical and research use.
  • Useful for both qualitative and quantitative hypoxia assessment.