Learn More
Medchemexpress LLC Pimonidazole hydrochloride, 50mg | CAS No. : 70132-51-3
SDP
Catalog No. NC3313365
Encompass_Preferred
Pimonidazole is a novel hypoxia marker for complementary study of tumor hypoxia and cell proliferation in tumor. Pimonidazole accumulates in hypoxic cells via covalent binding with macromolecules or by forming reductive metabolites after reduction of its nitro group, it can be used for qualitative and quantitative assessment of tumor hypoxia .
Catalog No. NC3313365 Supplier Medchemexpress LLC Supplier No. HY10512950MG
Pimonidazole is a novel hypoxia marker for complementary study of tumor hypoxia and cell proliferation in tumor. Pimonidazole accumulates in hypoxic cells via covalent binding with macromolecules or by forming reductive metabolites after reduction of its nitro group, it can be used for qualitative and quantitative assessment of tumor hypoxia .