Learn More
Medchemexpress LLC Pimonidazole hydrochloride | 70132-51-3 | MFCD01700599 | 100.0% | 290.75 | C11H19ClN4O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10512910MM 1ML
Encompass_Preferred
Pimonidazole hydrochloride is a nitroimidazole-derived hypoxia marker used to label and detect hypoxic cells in biological samples. It is supplied as a high-purity solid or as a pre-formulated aqueous solution for research applications, enabling studies of tumor hypoxia and cellular oxygenation.
  • Detects hypoxic cells via covalent binding under low-oxygen conditions.
  • Available as a 10 mM aqueous solution or as solid material for flexible workflows.
  • High purity supports reproducible experimental results.
  • Compatible with immunohistochemistry and imaging protocols.
  • Supplied in small-volume vials suitable for single-use applications.
Catalog No. 50-002-43654