Learn More
Medchemexpress LLC Pimonidazole (hydrochloride) | 70132-51-3 | MFCD01700599 | 100.0% | 290.75 | C11H19ClN4O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1051295MG
Encompass_Preferred
Pimonidazole hydrochloride is a nitroimidazole-based hypoxia marker used to label and detect hypoxic cells in tissues and cell cultures. It accumulates selectively in low-oxygen environments, enabling downstream detection by immunohistochemistry or biochemical assays. The product is supplied as a high-purity solid and should be stored sealed at recommended temperatures for stability.
  • Detects and accumulates in hypoxic cells for staining and quantification.
  • Suitable for immunohistochemistry, biochemical assays, and in vivo labeling.
  • High purity supports reproducible experimental results.
  • Available in small solid quantities appropriate for laboratory studies.
  • Stable when stored sealed at recommended temperatures (short-term and long-term conditions apply).
Catalog No. 50-002-43567