Learn More
Apexbio Technology LLC Pimozide 2062-78-4 250mg
SDP
Supplier:  Apexbio Technology LLC C3837250
Encompass_Preferred
Pimozide (CAS 2062-78-4) is a small-molecule antagonist targeting dopamine receptors including D2 D3 and D4 subtypes It is designed to inhibit dopamine receptor signaling thereby modulating dopaminergic pathways Pimozide exerts its biological activity primarily through antagonistic binding to dopamine receptors It demonstrates inhibition with Ki values of approximately 2 4 nM (D2) 0 2 nM (D3) and 1 8 nM (D4) In animal studies administration of pimozide reduced dopamine-driven behaviors such as food-motivated activity and cocaine reinforcement Based on these pharmacological properties pimozide holds research potential in neurological and psychiatric disorders including schizophrenia Tourette s syndrome dopamine-related motor disorders and delusional psychosis
Catalog No. 50-000-89423