Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pimozide-d4-1 | 00-00-0 | 465.57 g/mol | C28H25D4F2N3O | 5 MG
SDP

Catalog No. 5000263158
Encompass_Preferred

Pimozide-d4-1 is a deuterium-labeled isotopologue of pimozide supplied for research and analytical applications. The product is offered as a 5 mg quantity and is accompanied by a product data sheet and handling instructions for laboratory use.

  • Isotopically labeled (d4) for use as a mass-shifted internal standard.
  • Intended for research and analytical use.
  • Provided as a 5 mg powder.
  • Molecular formula C28H25D4F2N3O; molecular weight 465.57 g/mol.
  • Data sheet and handling instructions available for purity and handling information.

Catalog No. 50-002-63158 Supplier Medchemexpress LLC Supplier No. HY12987S25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pimozide-d4-1 is a deuterium-labeled isotopologue of pimozide supplied for research and analytical applications. The product is offered as a 5 mg quantity and is accompanied by a product data sheet and handling instructions for laboratory use.

  • Isotopically labeled (d4) for use as a mass-shifted internal standard.
  • Intended for research and analytical use.
  • Provided as a 5 mg powder.
  • Molecular formula C28H25D4F2N3O; molecular weight 465.57 g/mol.
  • Data sheet and handling instructions available for purity and handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.