Learn More
Medchemexpress LLC Pimozide-d4-1 | 00-00-0 | 98.0% | C28H25D4F2N3O | 10MG
SDP
Catalog No. 5000260004
Encompass_Preferred
Pimozide-d4-1 is the deuterium-labeled isotopologue of pimozide designed for use as an analytical tracer and internal standard in quantitative mass spectrometry and NMR methods. With four deuterium substitutions, the compound provides a distinct mass difference from the parent analyte to support accurate, interference-resistant measurement in bioanalytical and method-development workflows.
  • Enables accurate quantitation as an internal standard for LC-MS, GC-MS, or NMR.
  • Four deuterium labels provide a clear mass shift from the unlabeled analyte.
  • High purity supports reproducible analytical performance.
  • Suitable for method development and validation workflows.
  • Available in laboratory-scale pack sizes for routine use.
  • Supplied with documentation for analytical applications.
Catalog No. 50-002-60004 Supplier Medchemexpress LLC Supplier No. HY12987S210MG
Pimozide-d4-1 is the deuterium-labeled isotopologue of pimozide designed for use as an analytical tracer and internal standard in quantitative mass spectrometry and NMR methods. With four deuterium substitutions, the compound provides a distinct mass difference from the parent analyte to support accurate, interference-resistant measurement in bioanalytical and method-development workflows.
  • Enables accurate quantitation as an internal standard for LC-MS, GC-MS, or NMR.
  • Four deuterium labels provide a clear mass shift from the unlabeled analyte.
  • High purity supports reproducible analytical performance.
  • Suitable for method development and validation workflows.
  • Available in laboratory-scale pack sizes for routine use.
  • Supplied with documentation for analytical applications.