Learn More
Medchemexpress LLC Pimozide-d4-1 | 2062-78-4 | 1 MG
SDP
Catalog No. 5000340117
Encompass_Preferred
Pimozide-d4-1 is the deuterium labeled Pimozide. Pimozide is a dopamine receptor antagonist, with Kis of 1.4 nM, 2.5 nM and 588 nM for dopamine D2, D3 and D1 receptors, respectively, and also has affinity at α1-adrenoceptor, with a Ki of 39 nM. Additionally, Pimozide inhibits STAT3 and STAT5.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-40117 Supplier Medchemexpress LLC Supplier No. HY12987S21MG
Add to Cart
Add to Cart
Pimozide-d4-1 is the deuterium labeled Pimozide. Pimozide is a dopamine receptor antagonist, with Kis of 1.4 nM, 2.5 nM and 588 nM for dopamine D2, D3 and D1 receptors, respectively, and also has affinity at α1-adrenoceptor, with a Ki of 39 nM. Additionally, Pimozide inhibits STAT3 and STAT5.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.