Learn More
Medchemexpress LLC Pimozide-d4-1 | 2062-78-4 | 100 UG
SDP
Catalog No. 5000338525
Encompass_Preferred
Pimozide-d4-1 is a deuterium-labeled version of Pimozide, a dopamine receptor antagonist. It exhibits Ki values of 1.4 nM for D2, 2.5 nM for D3, and 588 nM for D1 receptors, along with affinity at the α1-adrenoceptor (Ki of 39 nM). Additionally, Pimozide inhibits STAT3 and STAT5. Deuterium labeling allows for its use as a tracer and internal standard in quantitative analyses, impacting pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Functions as a dopamine receptor antagonist
  • Exhibits affinity at α1-adrenoceptor
  • Inhibits STAT3 and STAT5
  • Useful as a tracer in drug development
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Impacts pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-38525 Supplier Medchemexpress LLC Supplier No. HY12987S2100UG
Pimozide-d4-1 is a deuterium-labeled version of Pimozide, a dopamine receptor antagonist. It exhibits Ki values of 1.4 nM for D2, 2.5 nM for D3, and 588 nM for D1 receptors, along with affinity at the α1-adrenoceptor (Ki of 39 nM). Additionally, Pimozide inhibits STAT3 and STAT5. Deuterium labeling allows for its use as a tracer and internal standard in quantitative analyses, impacting pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Functions as a dopamine receptor antagonist
  • Exhibits affinity at α1-adrenoceptor
  • Inhibits STAT3 and STAT5
  • Useful as a tracer in drug development
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Impacts pharmacokinetic and metabolic profiles of drugs