Learn More
Medchemexpress LLC Pimozide-d4 | 1803193-57-8 | 99.2% | 1 MG
SDP
Catalog No. 5000339094
Encompass_Preferred
Pimozide-d4 is a deuterium labeled Pimozide. It acts as a dopamine receptor antagonist with Ki values of 1.4 nM, 2.5 nM, and 588 nM for dopamine D2, D3, and D1 receptors, respectively. It also has an affinity at α1-adrenoceptor with a Ki of 39 nM, and inhibits STAT3 and STAT5. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Dopamine receptor antagonist (D2, D3, D1)
  • Affinity at α1-adrenoceptor
  • Inhibits STAT3 and STAT5
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-39094 Supplier Medchemexpress LLC Supplier No. HY12987S1MG
Pimozide-d4 is a deuterium labeled Pimozide. It acts as a dopamine receptor antagonist with Ki values of 1.4 nM, 2.5 nM, and 588 nM for dopamine D2, D3, and D1 receptors, respectively. It also has an affinity at α1-adrenoceptor with a Ki of 39 nM, and inhibits STAT3 and STAT5. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Dopamine receptor antagonist (D2, D3, D1)
  • Affinity at α1-adrenoceptor
  • Inhibits STAT3 and STAT5
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS