Learn More
Cayman Chemical Pimpinellin, Purity: ≥98%, CAS: 131-12-4, 50 mg
SDP
Catalog No. 502911539
Encompass_Preferred
A coumarin derivative with diverse biological activities; inhibits AChE and BChE by 23.54 and 66.55%, respectively, at 20 µg/ml; inhibits collagen-induced aggregation of mouse platelets ex vivo at 40 mg/kg; inhibits PTZ-induced seizure-like behavior in zebrafish larvae
Catalog No. 50-291-1539 Supplier Cayman Chemical Supplier No. 3627450 MG
A coumarin derivative with diverse biological activities; inhibits AChE and BChE by 23.54 and 66.55%, respectively, at 20 µg/ml; inhibits collagen-induced aggregation of mouse platelets ex vivo at 40 mg/kg; inhibits PTZ-induced seizure-like behavior in zebrafish larvae