Learn More
AdipoGen Pimprinine
SDP
Supplier:  AdipoGen BVT0297M005
Encompass_Preferred
Chemical. CAS 13640-26-1. Formula C12H10N2O. MW 198.2. Isolated from Streptomyces sp. Indole alkaloid. Monoamine oxidase inhibitor MAOI. Antiepileptic anticonvulsant compound. Inhibitor of platelet aggregation and thromboxane A2 synthesis. Weak antifungal and antituberculosis activity. Useful as lead structure for potent antifungal substances.
Catalog No. 50-256-2007