Learn More
Med Vet International Pin Cutter, J0220
Catalog No. 5000584806
Encompass_Preferred
For cutting intramedullary pins and customizing bone plates. Stainless steel. 1470g. 48cm (19"L). Cuts up to 6.4mm (1/4") pins. Narrow jaws allow extra-close pin cutting
Catalog No. 50-005-84806 Supplier Med Vet International Supplier No. JOR220
For cutting intramedullary pins and customizing bone plates. Stainless steel. 1470g. 48cm (19"L). Cuts up to 6.4mm (1/4") pins. Narrow jaws allow extra-close pin cutting