Learn More
Grainger Pin Gage .0050 Plus Class ZZ
Catalog No. 502749196
Encompass_Preferred
The Vermont Gage 140G84 pin gage is a precision measuring instrument used to ensure accurate size and tolerance of holes, shafts, slots, and other components. It has a diameter of 0.1405 inches, a tolerance of +/- 0.00005 inches, and is made of hardened and ground tool steel for durability and accuracy. The gage has a knurled grip, carbide measuring surfaces, and includes a plastic storage case. It can be used for quality control inspection and is suitable for workshop, laboratory, and production use. The pin gage allows precise measurement and verification of the size of openings and parts.
Catalog No. 50-274-9196 Supplier Grainger Supplier No. 140G84
The Vermont Gage 140G84 pin gage is a precision measuring instrument used to ensure accurate size and tolerance of holes, shafts, slots, and other components. It has a diameter of 0.1405 inches, a tolerance of +/- 0.00005 inches, and is made of hardened and ground tool steel for durability and accuracy. The gage has a knurled grip, carbide measuring surfaces, and includes a plastic storage case. It can be used for quality control inspection and is suitable for workshop, laboratory, and production use. The pin gage allows precise measurement and verification of the size of openings and parts.