Learn More
Grainger Pin Gage .0110 Plus Class ZZ
Catalog No. 502753534
Encompass_Preferred
The Westward Pin Gage Set is a precision measuring tool used to check hole sizes and verify part dimensions. It contains 13 pin gages with diameters ranging from 0.228" to 0.244" in 0.001" increments. The gages are made from hardened and ground tool steel for durability and accuracy. They feature a knurled grip area for easy handling. This pin gage set allows for quick go/no-go checking and is suitable for use in production, inspection, and engineering applications. The set comes packaged in a plastic case for storage and organization.
Catalog No. 50-275-3534 Supplier Grainger Supplier No. 140G90
The Westward Pin Gage Set is a precision measuring tool used to check hole sizes and verify part dimensions. It contains 13 pin gages with diameters ranging from 0.228" to 0.244" in 0.001" increments. The gages are made from hardened and ground tool steel for durability and accuracy. They feature a knurled grip area for easy handling. This pin gage set allows for quick go/no-go checking and is suitable for use in production, inspection, and engineering applications. The set comes packaged in a plastic case for storage and organization.