Learn More
Grainger Pin Gage .0190 Plus Class ZZ
Catalog No. 502753420
Encompass_Preferred
The Westward Pin Gage Set is a precision measuring tool used to check hole sizes and verify part dimensions. It contains 13 pin gages with diameters ranging from 0.228" to 0.244" in 0.001" increments. The gages are made from heat treated and hardened alloy steel for durability and accuracy. They feature a knurled grip area for easy handling. This pin gage set allows for quick go/no-go checking and is suitable for use in production, inspection, and engineering applications. The gages come packed in a fitted plastic case for storage and organization.
Catalog No. 50-275-3420 Supplier Grainger Supplier No. 140G98
The Westward Pin Gage Set is a precision measuring tool used to check hole sizes and verify part dimensions. It contains 13 pin gages with diameters ranging from 0.228" to 0.244" in 0.001" increments. The gages are made from heat treated and hardened alloy steel for durability and accuracy. They feature a knurled grip area for easy handling. This pin gage set allows for quick go/no-go checking and is suitable for use in production, inspection, and engineering applications. The gages come packed in a fitted plastic case for storage and organization.