Learn More
Grainger Pin Gage .0340 Plus Class ZZ
Catalog No. 502750030
Encompass_Preferred
The Vermont Gage Pin Gage 140H13 is a precision measuring instrument used to check the size and tolerance of holes and shafts. It is made from hardened and tempered tool steel to ensure durability and accuracy. This pin gage has a diameter of 0.140 inches, a tolerance of +/- 0.00005 inches, and an included angle of 30 degrees. It can be used to perform quick go/no-go checks on parts and is suitable for workshop, inspection room, and production line use. The knurled grip on the handle provides a secure hold during measurement. This pin gage is manufactured in the USA and calibrated to NIST standards for reliable and repeatable measurements.
Catalog No. 50-275-0030 Supplier Grainger Supplier No. 140H13
The Vermont Gage Pin Gage 140H13 is a precision measuring instrument used to check the size and tolerance of holes and shafts. It is made from hardened and tempered tool steel to ensure durability and accuracy. This pin gage has a diameter of 0.140 inches, a tolerance of +/- 0.00005 inches, and an included angle of 30 degrees. It can be used to perform quick go/no-go checks on parts and is suitable for workshop, inspection room, and production line use. The knurled grip on the handle provides a secure hold during measurement. This pin gage is manufactured in the USA and calibrated to NIST standards for reliable and repeatable measurements.