Learn More
Grainger Pin Gage .0520 Plus Class ZZ
Catalog No. 502749195
Encompass_Preferred
The Grainger plug and pin gauge sets are precision measuring tools used to check hole sizes and shaft diameters. They come in a wide range of imperial and metric sizes. The plug gauges have a go/no-go tolerance to accurately check hole diameters. The pin gauges allow precise measurement of shaft diameters. The sets are made of durable hardened steel and come in a protective case. Common sizes are fractional, number, and letter drill sizes. They are an essential inspection tool for any machine shop or manufacturer working with precision holes or shafts.
Catalog No. 50-274-9195 Supplier Grainger Supplier No. 140H31
The Grainger plug and pin gauge sets are precision measuring tools used to check hole sizes and shaft diameters. They come in a wide range of imperial and metric sizes. The plug gauges have a go/no-go tolerance to accurately check hole diameters. The pin gauges allow precise measurement of shaft diameters. The sets are made of durable hardened steel and come in a protective case. Common sizes are fractional, number, and letter drill sizes. They are an essential inspection tool for any machine shop or manufacturer working with precision holes or shafts.