Learn More
Grainger Pin Gage .0600 Plus Class ZZ
Catalog No. 502749039
Encompass_Preferred
The Grainger Approved Pin Gage is a precision measuring tool used to check the size of holes, slots, grooves and other internal dimensions. It has a hardened and ground steel pin that is 0.140 inches in diameter with a tolerance of +/- 0.00005 inches. The pin is knurled for easy handling and removal. The gage has a satin chrome finish to resist corrosion. It can measure hole sizes between 0.1395 and 0.1405 inches. The gage is made in the USA and conforms to ASME B89.1.6 standards. The Grainger Approved brand offers quality tools at a great value. This durable pin gage is ideal for use in production, inspection and engineering applications.
Catalog No. 50-274-9039 Supplier Grainger Supplier No. 140H39
The Grainger Approved Pin Gage is a precision measuring tool used to check the size of holes, slots, grooves and other internal dimensions. It has a hardened and ground steel pin that is 0.140 inches in diameter with a tolerance of +/- 0.00005 inches. The pin is knurled for easy handling and removal. The gage has a satin chrome finish to resist corrosion. It can measure hole sizes between 0.1395 and 0.1405 inches. The gage is made in the USA and conforms to ASME B89.1.6 standards. The Grainger Approved brand offers quality tools at a great value. This durable pin gage is ideal for use in production, inspection and engineering applications.