Learn More
Grainger Pin Gage .0960 Plus Class ZZ
Supplier:  Grainger 140H74
Encompass_Preferred
The Vermont Gage 140H74 pin gage is a precision measuring instrument used to accurately measure the size of holes, slots, or other internal dimensions. It has a diameter of 0.1405 inches, a tolerance of +/- 0.00005 inches, and is made from hardened and ground alloy steel. The gage has a knurled grip, carbide measuring surfaces, and includes a plastic case. It can be used to check tolerances, inspect parts, or validate that dimensions meet specifications. The high precision of the gage makes it well-suited for use in industries like aerospace, automotive, and manufacturing where tight tolerances are required.
Catalog No. 50-275-1006