Learn More
Grainger Pin Gage .1040 Plus Class ZZ
Catalog No. 502750768
Encompass_Preferred
The Grainger Approved Pin Gage is a precision tool for measuring holes, slots, and grooves. It features a 0.140-inch diameter pin with a tolerance of +/- 0.00005 inches, made from heat-treated steel with a matte chrome finish. The knurled grip ensures comfort and control. Ideal for industrial and engineering applications, it serves as a reference standard for setting micrometers. The gage includes a plastic case for protection, making it a high-quality choice for precise measurements.
Catalog No. 50-275-0768 Supplier Grainger Supplier No. 140H82
The Grainger Approved Pin Gage is a precision tool for measuring holes, slots, and grooves. It features a 0.140-inch diameter pin with a tolerance of +/- 0.00005 inches, made from heat-treated steel with a matte chrome finish. The knurled grip ensures comfort and control. Ideal for industrial and engineering applications, it serves as a reference standard for setting micrometers. The gage includes a plastic case for protection, making it a high-quality choice for precise measurements.