Learn More
Grainger Pin Gage .1150 Plus Class ZZ
Catalog No. 502751008
Encompass_Preferred
The Vermont Gage Pin Gage 140H13 is a precision measuring instrument used to check the size and tolerance of holes, shafts, slots, and other cylindrical features. It is made from hardened tool steel and ground to a tolerance of +/- .0001". The pin diameter is .140" with a carbide tip and knurled grip. It has a measuring range of .1395" - .1405". This pin gage allows for quick go/no-go checking and is suitable for workshop, inspection room, and production use. The knurled grip provides a comfortable, slip-resistant handling. It meets federal specification GGG-P-463C and is serialized for traceability. This is a high precision pin gage for accurate measurement and quality control.
Catalog No. 50-275-1008 Supplier Grainger Supplier No. 140H93
The Vermont Gage Pin Gage 140H13 is a precision measuring instrument used to check the size and tolerance of holes, shafts, slots, and other cylindrical features. It is made from hardened tool steel and ground to a tolerance of +/- .0001". The pin diameter is .140" with a carbide tip and knurled grip. It has a measuring range of .1395" - .1405". This pin gage allows for quick go/no-go checking and is suitable for workshop, inspection room, and production use. The knurled grip provides a comfortable, slip-resistant handling. It meets federal specification GGG-P-463C and is serialized for traceability. This is a high precision pin gage for accurate measurement and quality control.